The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100819.2 | NP_001094289.1 | 5 - 6 | 416 | 89 |
NM_001204094.1 | NP_001191023.1 | 6 - 7 | 433 | 89 | |
NM_015387.4 | NP_056202.2 | 6 - 7 | 479 | 89 | |
NM_199482.3 | NP_955776.1 | 6 - 7 | 489 | 89 | |
GenBank mRNA | AB015441.1 | - | 6 - 7 | 438 | 89 |
AF093825.1 | - | 6 - 7 | 454 | 89 | |
AF151853.1 | - | 6 - 7 | 454 | 89 | |
AF250319.1 | - | 6 - 7 | 464 | 89 | |
AJ580638.1 | - | 6 - 7 | 435 | 89 | |
AK027043.1 | - | 6 - 7 | 476 | 89 | |
AK289377.1 | - | 6 - 7 | 469 | 89 | |
AK292938.1 | - | 6 - 7 | 475 | 89 | |
AK297294.1 | - | - | 350 | 89 | |
AK297514.1 | - | 5 - 6 | 375 | 89 | |
AK307497.1 | - | 5 - 6 | 347 | 89 | |
BC005237.1 | - | 6 - 7 | 479 | 89 | |
CR457371.1 | - | 6 - 7 | 435 | 89 | |
DC298610.1 | - | 6 - 7 | 489 | 89 | |
DC350915.1 | - | 5 - 6 | 412 | 89 | |
DC375649.1 | - | 6 - 7 | 435 | 89 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202485.1 | NP_001189414.1 | 7 - 8 | 983 | 89 |