The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321357.1 | NP_001308286.1 | 17 - 18 | 2276 | 59 |
NM_001321358.1 | NP_001308287.1 | 17 - 18 | 1673 | 59 | |
NM_001321359.1 | NP_001308288.1 | 17 - 18 | 1668 | 59 | |
NM_001321361.1 | NP_001308290.1 | 17 - 18 | 1670 | 59 | |
NM_001321362.1 | NP_001308291.1 | 16 - 17 | 1646 | 59 | |
NM_001321363.1 | NP_001308292.1 | 16 - 17 | 1553 | 59 | |
NM_013392.3 | NP_037524.1 | 16 - 17 | 1644 | 59 | |
GenBank mRNA | AF113249.1 | - | 16 - 17 | 1556 | 59 |
AK001946.1 | - | 16 - 17 | 1534 | 59 | |
AK027538.1 | - | 16 - 17 | 1552 | 59 | |
AK092860.1 | - | 9 - 10 | 1954 | 59 | |
AK122664.1 | - | 17 - 18 | 2276 | 59 | |
AK130619.1 | - | 9 - 10 | 688 | 59 | |
AK223136.1 | - | 16 - 17 | 1540 | 59 | |
AK301345.1 | - | 17 - 18 | 1487 | 59 | |
AL136682.1 | - | 16 - 17 | 1531 | 59 | |
BC001221.1 | - | 16 - 17 | 1558 | 59 | |
BQ225378.1 | - | 2 - 3 | 93 | 59 |