Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
Chromosome Location
Chr.6: 47477746 - 47627263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012120.2 NP_036252.1 13 - 14 1879 108
XM_005248976.1 XP_005249033.1 13 - 14 1910 108
XM_011514449.2 XP_011512751.1 12 - 13 1501 108
GenBank mRNA AF146277.1 - 13 - 14 1473 108
AF164377.1 - 12 - 13 1423 108
BC069444.1 - 13 - 14 1469 108

View Full Product Details