Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
Chromosome Location
Chr.6: 47477746 - 47627263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012120.2 NP_036252.1 4 - 5 881 86
XM_005248976.1 XP_005249033.1 4 - 5 924 86
XM_011514449.2 XP_011512751.1 3 - 4 503 86
XM_017010641.1 XP_016866130.1 4 - 5 924 86
GenBank mRNA AF146277.1 - 4 - 5 475 86
AF164377.1 - 3 - 4 425 86
BC069444.1 - 4 - 5 471 86

View Full Product Details