The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282225.1 | NP_001269154.1 | 4 - 5 | 1029 | 68 |
NM_001282226.1 | NP_001269155.1 | 4 - 5 | 996 | 68 | |
NM_001282227.1 | NP_001269156.1 | 4 - 5 | 802 | 68 | |
NM_001282228.1 | NP_001269157.1 | 4 - 5 | 904 | 68 | |
NM_001282229.1 | NP_001269158.1 | 3 - 4 | 661 | 68 | |
XM_006724080.3 | XP_006724143.1 | 4 - 5 | 616 | 68 | |
XM_011546133.1 | XP_011544435.1 | 4 - 5 | 921 | 68 | |
GenBank mRNA | AB209648.1 | - | 4 - 5 | 616 | 68 |
AF190746.1 | - | 3 - 4 | 965 | 68 | |
AK292689.1 | - | 4 - 5 | 1017 | 68 | |
AK295174.1 | - | 4 - 5 | 753 | 68 | |
AK304818.1 | - | 4 - 5 | 796 | 68 | |
AK314321.1 | - | 4 - 5 | 820 | 68 | |
BC051755.1 | - | 3 - 4 | 800 | 68 | |
CR456417.1 | - | 3 - 4 | 787 | 68 |