The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282225.1 | NP_001269154.1 | 8 - 9 | 1516 | 64 |
NM_001282226.1 | NP_001269155.1 | 8 - 9 | 1483 | 64 | |
NM_001282227.1 | NP_001269156.1 | 8 - 9 | 1289 | 64 | |
NM_001282228.1 | NP_001269157.1 | 8 - 9 | 1391 | 64 | |
NM_001282229.1 | NP_001269158.1 | 7 - 8 | 1148 | 64 | |
NM_177405.2 | NP_803124.1 | 5 - 6 | 722 | 64 | |
XM_006724080.3 | XP_006724143.1 | 7 - 8 | 2753 | 64 | |
XM_011546133.1 | XP_011544435.1 | 8 - 9 | 1408 | 64 | |
GenBank mRNA | AB209648.1 | - | 7 - 8 | 2753 | 64 |
AF190746.1 | - | 7 - 8 | 1452 | 64 | |
AK027682.1 | - | 5 - 6 | 543 | 64 | |
AK074702.1 | - | 5 - 6 | 594 | 64 | |
AK130725.1 | - | 2 - 3 | 236 | 64 | |
AK292689.1 | - | 8 - 9 | 1504 | 64 | |
AK304818.1 | - | 8 - 9 | 1283 | 64 | |
AK314321.1 | - | 8 - 9 | 1307 | 64 | |
BC051755.1 | - | 7 - 8 | 1287 | 64 | |
CR456417.1 | - | 7 - 8 | 1274 | 64 |