The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017829.5 | NP_060299.4 | 2 - 3 | 280 | 70 |
NM_033070.2 | NP_149061.1 | 2 - 3 | 355 | 70 | |
XM_005261236.3 | XP_005261293.1 | 2 - 3 | 281 | 70 | |
XM_011546127.1 | XP_011544429.1 | 2 - 3 | 770 | 70 | |
XM_017028781.1 | XP_016884270.1 | 2 - 3 | 770 | 70 | |
XM_017028782.1 | XP_016884271.1 | 2 - 3 | 333 | 70 | |
XM_017028783.1 | XP_016884272.1 | 2 - 3 | 770 | 70 | |
XM_017028784.1 | XP_016884273.1 | 2 - 3 | 281 | 70 | |
GenBank mRNA | AF273270.1 | - | 2 - 3 | 280 | 70 |
AF273271.1 | - | 2 - 3 | 340 | 70 | |
AK000461.1 | - | 2 - 3 | 353 | 70 | |
AK001034.1 | - | 2 - 3 | 333 | 70 | |
AK315156.1 | - | 2 - 3 | 341 | 70 | |
BC042540.1 | - | 2 - 3 | 339 | 70 |