The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020943.2 | NP_065994.1 | 11 - 12 | 1504 | 135 |
XM_005246726.2 | XP_005246783.1 | 11 - 12 | 1607 | 135 | |
GenBank mRNA | AB046824.1 | - | 10 - 11 | 1323 | 135 |
AK025635.1 | - | 11 - 12 | 1361 | 135 | |
AK026978.1 | - | 4 - 5 | 461 | 135 | |
BC016651.1 | - | 10 - 11 | 1328 | 135 | |
BC031216.1 | - | 11 - 12 | 1504 | 135 | |
BC041662.1 | - | 4 - 5 | 455 | 135 | |
BC053573.1 | - | 8 - 9 | 1026 | 135 | |
BC057826.1 | - | 11 - 12 | 1362 | 135 | |
BC093952.1 | - | 10 - 11 | 1226 | 135 | |
BC093954.1 | - | 10 - 11 | 1226 | 135 | |
BC143433.1 | - | 9 - 10 | 1309 | 135 |