The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020943.2 | NP_065994.1 | 17 - 18 | 2114 | 120 |
XM_005246726.2 | XP_005246783.1 | 17 - 18 | 2217 | 120 | |
GenBank mRNA | AB046824.1 | - | 16 - 17 | 1933 | 120 |
AK025635.1 | - | 17 - 18 | 1970 | 120 | |
AK026978.1 | - | 10 - 11 | 1071 | 120 | |
AK123647.1 | - | 4 - 5 | 1071 | 120 | |
BC016651.1 | - | 16 - 17 | 1938 | 120 | |
BC031216.1 | - | 17 - 18 | 2114 | 120 | |
BC041662.1 | - | 10 - 11 | 1065 | 120 | |
BC053573.1 | - | 14 - 15 | 1636 | 120 | |
BC057826.1 | - | 17 - 18 | 1972 | 120 | |
BC093952.1 | - | 16 - 17 | 1836 | 120 | |
BC093954.1 | - | 16 - 17 | 1836 | 120 |