Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSNK1G3
Entrez Gene ID
1456
Gene Name
casein kinase 1 gamma 3
Gene Aliases
CKI-gamma 3, CSNK1G3L
Chromosome Location
Chr.5: 123512099 - 123617045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031812.3 NP_001026982.1 5 - 6 1160 150
NM_001044723.2 NP_001038188.1 4 - 5 688 150
NM_001270572.1 NP_001257501.1 4 - 5 688 150
NM_001270573.1 NP_001257502.1 4 - 5 735 150
NM_001270574.1 NP_001257503.1 2 - 3 624 150
NM_004384.4 NP_004375.2 4 - 5 688 150
XM_005271891.1 XP_005271948.1 5 - 6 1160 150
XM_005271892.2 XP_005271949.1 5 - 6 1160 150
XM_005271894.2 XP_005271951.1 5 - 6 1160 150
XM_005271895.2 XP_005271952.1 5 - 6 1160 150
XM_005271896.1 XP_005271953.1 5 - 6 1160 150
XM_017009057.1 XP_016864546.1 6 - 7 1235 150
XM_017009058.1 XP_016864547.1 8 - 9 1444 150
XM_017009059.1 XP_016864548.1 7 - 8 1303 150
XM_017009060.1 XP_016864549.1 7 - 8 1359 150
XM_017009061.1 XP_016864550.1 6 - 7 1235 150
XM_017009062.1 XP_016864551.1 6 - 7 794 150
XM_017009063.1 XP_016864552.1 6 - 7 1235 150
XM_017009064.1 XP_016864553.1 6 - 7 1235 150
XM_017009065.1 XP_016864554.1 6 - 7 1235 150
XM_017009066.1 XP_016864555.1 7 - 8 1303 150
XM_017009067.1 XP_016864556.1 6 - 7 1235 150
XM_017009068.1 XP_016864557.1 6 - 7 1235 150
XM_017009069.1 XP_016864558.1 6 - 7 1228 150
XM_017009070.1 XP_016864559.1 5 - 6 1160 150
GenBank mRNA AK289405.1 - 4 - 5 633 150
AK299739.1 - 4 - 5 574 150
AK316019.1 - 2 - 3 463 150
BC047567.1 - 5 - 6 812 150

View Full Product Details