The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198850.1 | NP_001185779.1 | 2 - 3 | 316 | 79 |
NM_001198851.1 | NP_001185780.1 | 4 - 5 | 359 | 79 | |
NM_001198852.1 | NP_001185781.1 | 2 - 3 | 221 | 79 | |
NM_014947.4 | NP_055762.3 | 2 - 3 | 238 | 79 | |
XM_011541026.2 | XP_011539328.1 | 3 - 4 | 290 | 79 | |
GenBank mRNA | AB028964.1 | - | 4 - 5 | 359 | 79 |
AK027075.1 | - | 2 - 3 | 316 | 79 | |
BC014182.2 | - | 2 - 3 | 221 | 79 | |
BC151828.1 | - | 2 - 3 | 207 | 79 | |
BC152441.1 | - | 4 - 5 | 359 | 79 |