The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003643.3 | NP_003634.2 | 4 - 5 | 652 | 78 |
XM_017011390.1 | XP_016866879.1 | 8 - 9 | 2034 | 78 | |
GenBank mRNA | AB026493.1 | - | 3 - 4 | 523 | 78 |
AB041714.1 | - | 3 - 4 | 443 | 78 | |
BC096288.1 | - | 4 - 5 | 610 | 78 | |
D88613.1 | - | 4 - 5 | 655 | 78 |