The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000572.2 | NP_000563.1 | 2 - 3 | 282 | 138 |
GenBank mRNA | AF043333.1 | - | 2 - 3 | 169 | 138 |
AY029171.1 | - | 2 - 3 | 223 | 138 | |
BC104252.1 | - | 2 - 3 | 279 | 138 | |
BC104253.1 | - | 2 - 3 | 279 | 138 | |
CR541993.1 | - | 2 - 3 | 223 | 138 | |
CR542028.1 | - | 2 - 3 | 223 | 138 | |
M57627.1 | - | 2 - 3 | 253 | 138 |