Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABBR1
Entrez Gene ID
2550
Gene Name
gamma-aminobutyric acid type B receptor subunit 1
Gene Aliases
GABABR1, GABBR1-3, GB1, GPRC3A
Chromosome Location
Chr.6: 29602228 - 29633135 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319053.1 NP_001305982.1 14 - 15 1927 89
NM_001470.3 NP_001461.1 14 - 15 1991 89
NM_021903.2 NP_068703.1 9 - 10 1791 89
NM_021904.3 NP_068704.2 13 - 14 1805 89
XM_005248982.2 XP_005249039.1 14 - 15 1806 89
XM_006715047.3 XP_006715110.1 13 - 14 1606 89
XM_011514453.2 XP_011512755.1 12 - 13 1585 89
XM_011514455.1 XP_011512757.1 14 - 15 1806 89
XM_017010676.1 XP_016866165.1 10 - 11 1337 89
GenBank mRNA AB208790.1 - 14 - 15 1726 89
AF099148.1 - 14 - 15 1813 89
AJ012185.1 - 13 - 14 1705 89
AJ012186.1 - 9 - 10 1354 89
AJ012187.1 - 12 - 13 1519 89
AJ012288.1 - 14 - 15 1748 89
AJ225028.1 - 14 - 15 1939 89
AJ225029.1 - 9 - 10 1533 89
AK223619.1 - 9 - 10 1576 89
AK290191.1 - 9 - 10 1576 89
AK312308.1 - 9 - 10 1572 89
BC041332.2 - 9 - 10 1576 89
BC042598.1 - 9 - 10 1565 89
BC050532.1 - 9 - 10 1563 89
Y11044.1 - 13 - 14 1702 89

View Full Product Details