The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001255.2 | NP_001246.2 | 8 - 9 | 1179 | 109 |
GenBank mRNA | AF099644.1 | - | 7 - 8 | 1122 | 109 |
AK225504.1 | - | 8 - 9 | 109 | ||
AK303844.1 | - | 9 - 10 | 1134 | 109 | |
AK312780.1 | - | 8 - 9 | 1158 | 109 | |
BC000624.2 | - | 7 - 8 | 1186 | 109 | |
BC001088.2 | - | 8 - 9 | 1179 | 109 | |
BC006272.2 | - | 7 - 8 | 1121 | 109 | |
BC009425.1 | - | 8 - 9 | 1161 | 109 | |
BC009426.1 | - | 8 - 9 | 1161 | 109 | |
BC010044.2 | - | 8 - 9 | 1170 | 109 | |
BC012803.2 | - | 8 - 9 | 1161 | 109 | |
BC012827.2 | - | 8 - 9 | 1179 | 109 | |
BC013303.2 | - | 8 - 9 | 1161 | 109 | |
BC015998.1 | - | 8 - 9 | 1157 | 109 | |
BC024257.1 | - | 8 - 9 | 1161 | 109 | |
BC031294.1 | - | 8 - 9 | 1161 | 109 | |
BC110321.1 | - | 7 - 8 | 1142 | 109 | |
BT007388.1 | - | 7 - 8 | 1078 | 109 | |
U05340.1 | - | 8 - 9 | 1188 | 109 |