The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021080.3 | NP_066566.3 | 8 - 9 | 1232 | 83 |
GenBank mRNA | AB210012.1 | - | 8 - 9 | 1214 | 83 |
AF071062.1 | - | 5 - 6 | 558 | 83 | |
AK095513.1 | - | 8 - 9 | 1232 | 83 | |
AK313144.1 | - | 6 - 7 | 879 | 83 |