Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM10
Entrez Gene ID
10107
Gene Name
tripartite motif containing 10
Gene Aliases
HERF1, RFB30, RNF9
Chromosome Location
Chr.6: 30151939 - 30163216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006778.3 NP_006769.2 1 - 2 508 152
NM_052828.2 NP_439893.2 1 - 2 508 152
XM_011514221.1 XP_011512523.1 3 - 4 756 152
XM_011514222.2 XP_011512524.1 3 - 4 990 152
XM_011514223.2 XP_011512525.1 4 - 5 871 152
XM_011514225.1 XP_011512527.1 3 - 4 756 152
GenBank mRNA AF220122.1 - 1 - 2 506 152
AF220123.1 - 1 - 2 506 152
BC093926.1 - 1 - 2 486 152
BC113419.1 - 1 - 2 486 152

View Full Product Details