Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSG1
Entrez Gene ID
79363
Gene Name
REM2 and RAB like small GTPase 1
Gene Aliases
C1orf89
Chromosome Location
Chr.1: 16231687 - 16237445 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030907.3 NP_112169.2 4 - 5 947 58
GenBank mRNA BC002946.2 - 4 - 5 734 58
BC008702.1 - 4 - 5 734 58

View Full Product Details