The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042519.1 | NP_001035984.1 | 1 - 2 | 246 | 142 |
GenBank mRNA | DA247977.1 | - | 1 - 2 | 246 | 142 |