The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283012.1 | NP_001269941.1 | 5 - 6 | 886 | 73 |
NM_022783.3 | NP_073620.2 | 7 - 8 | 1189 | 73 | |
XM_017013735.1 | XP_016869224.1 | 5 - 6 | 632 | 73 | |
GenBank mRNA | AK022490.1 | - | 7 - 8 | 1077 | 73 |
AK023916.1 | - | 7 - 8 | 973 | 73 | |
AK225168.1 | - | 7 - 8 | 1122 | 73 | |
AK297822.1 | - | 5 - 6 | 819 | 73 | |
AK315175.1 | - | 7 - 8 | 1127 | 73 | |
AL136678.1 | - | 7 - 8 | 1119 | 73 | |
BC012040.1 | - | 7 - 8 | 1033 | 73 | |
BC024746.1 | - | 7 - 8 | 1089 | 73 |