Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP19-1
Entrez Gene ID
337882
Gene Name
keratin associated protein 19-1
Gene Aliases
KAP19.1
Chromosome Location
Chr.21: 30480045 - 30480317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ457067.1 - 1 - 1 465 125

View Full Product Details