Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
Chromosome Location
Chr.1: 34166883 - 34219131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134734.1 NP_001128206.1 6 - 7 2556 57
NM_032884.4 NP_116273.2 6 - 7 1687 57
GenBank mRNA AA725646.1 - 1 - 2 57
AK123355.1 - 6 - 7 1687 57
BC007637.1 - 6 - 7 1455 57
BC064845.1 - 6 - 7 1838 57

View Full Product Details