The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271995.1 | NP_001258924.1 | 1 - 2 | 180 | 91 |
NM_001271996.1 | NP_001258925.1 | 1 - 2 | 180 | 91 | |
NM_001271997.1 | NP_001258926.1 | 1 - 2 | 180 | 91 | |
NM_032607.2 | NP_115996.1 | 1 - 2 | 180 | 91 | |
GenBank mRNA | AB050902.1 | - | 1 - 2 | 149 | 91 |
AB073612.1 | - | 1 - 2 | 141 | 91 | |
AK172839.1 | - | 1 - 2 | 149 | 91 | |
AK313099.1 | - | 1 - 2 | 152 | 91 | |
BC101504.1 | - | 1 - 2 | 113 | 91 | |
BC101508.1 | - | 1 - 2 | 109 | 91 | |
BC143609.1 | - | 1 - 2 | 113 | 91 | |
BC143610.1 | - | 1 - 2 | 113 | 91 |