Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DISC1
- Entrez Gene ID
-
27185
- Gene Name
- disrupted in schizophrenia 1
- Gene Aliases
-
C1orf136, SCZD9
- Chromosome Location
-
Chr.1: 231626815 - 232041272 on Build GRCh38
- Gene Symbol
- TSNAX-DISC1
- Entrez Gene ID
-
100303453
- Gene Name
- TSNAX-DISC1 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 231528653 - 232041272 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028393.1 |
|
13 - 14 |
2708 |
103 |
View Full Product Details