Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSX
Entrez Gene ID
390259
Gene Name
brain specific homeobox
Gene Aliases
BSX1
Chromosome Location
Chr.11: 122977649 - 122981671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098169.1 NP_001091639.1 2 - 3 465 87

View Full Product Details