Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FXYD3
Entrez Gene ID
5349
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT8, PLML
Chromosome Location
Chr.19: 35115821 - 35124324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136008.1 NP_001129480.1 2 - 3 229 56
NM_001136010.1 NP_001129482.1 2 - 3 229 56
NM_001136012.1 NP_001129484.1 2 - 3 229 56
NM_005971.3 NP_005962.1 2 - 3 229 56
GenBank mRNA BC005238.1 - 1 - 2 43 56
BC090044.1 - 2 - 3 178 56
DA438143.1 - 1 - 2 43 56
DA890604.1 - 1 - 2 71 56
DB202948.1 - 1 - 2 44 56
X93036.1 - 1 - 2 46 56

View Full Product Details