The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000186.3 | NP_000177.2 | 3 - 4 | 592 | 109 |
NM_001014975.2 | NP_001014975.1 | 3 - 4 | 592 | 109 | |
XM_017001108.1 | XP_016856597.1 | 3 - 4 | 474 | 109 | |
GenBank mRNA | AK225649.1 | - | 3 - 4 | 432 | 109 |
AK291395.1 | - | 3 - 4 | 431 | 109 | |
BC012610.1 | - | 3 - 4 | 418 | 109 | |
BC037285.2 | - | 3 - 4 | 417 | 109 | |
BC110643.1 | - | 3 - 4 | 408 | 109 | |
BC142699.1 | - | 3 - 4 | 404 | 109 | |
X04697.1 | - | 2 - 3 | 196 | 109 | |
X07523.1 | - | 3 - 4 | 425 | 109 | |
Y00716.1 | - | 3 - 4 | 425 | 109 |