Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFH
Entrez Gene ID
3075
Gene Name
complement factor H
Gene Aliases
AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS
Chromosome Location
Chr.1: 196651878 - 196747504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000186.3 NP_000177.2 5 - 6 857 178
NM_001014975.2 NP_001014975.1 5 - 6 857 178
XM_017001108.1 XP_016856597.1 5 - 6 739 178
GenBank mRNA AK225649.1 - 5 - 6 697 178
AK291395.1 - 5 - 6 696 178
BC037285.2 - 5 - 6 682 178
BC110643.1 - 5 - 6 673 178
BC142699.1 - 5 - 6 669 178
X04697.1 - 4 - 5 461 178
X07523.1 - 5 - 6 690 178
Y00716.1 - 5 - 6 690 178

View Full Product Details