The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172439.1 | NP_001165910.1 | 5 - 6 | 710 | 78 |
NM_001172440.1 | NP_001165911.1 | 3 - 4 | 521 | 78 | |
NM_006025.3 | NP_006016.1 | 4 - 5 | 587 | 78 | |
GenBank mRNA | AK075446.1 | - | 5 - 6 | 674 | 78 |
AK295963.1 | - | 2 - 3 | 309 | 78 | |
AK300169.1 | - | 3 - 4 | 483 | 78 | |
AK302308.1 | - | 2 - 3 | 301 | 78 | |
AK314688.1 | - | 4 - 5 | 457 | 78 | |
BC069715.1 | - | 4 - 5 | 466 | 78 | |
BC074763.2 | - | 4 - 5 | 472 | 78 | |
BC111782.1 | - | 4 - 5 | 561 | 78 | |
M32402.1 | - | 4 - 5 | 582 | 78 | |
M36109.1 | - | 4 - 5 | 582 | 78 |