The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019016.2 | NP_061889.2 | 5 - 6 | 1199 | 82 |
XM_006721739.3 | XP_006721802.1 | 6 - 7 | 890 | 82 | |
XM_011524460.2 | XP_011522762.1 | 6 - 7 | 875 | 82 | |
XM_017024299.1 | XP_016879788.1 | 5 - 6 | 1184 | 82 | |
GenBank mRNA | AK000268.1 | - | 5 - 6 | 1199 | 82 |
BC111968.1 | - | 5 - 6 | 1174 | 82 |