The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114092.1 | NP_001107564.1 | 2 - 3 | 401 | 116 |
NM_015453.2 | NP_056268.2 | 2 - 3 | 606 | 116 | |
XM_005265021.4 | XP_005265078.1 | 2 - 3 | 963 | 116 | |
XM_005265022.4 | XP_005265079.1 | 2 - 3 | 1166 | 116 | |
XM_005265023.3 | XP_005265080.1 | 2 - 3 | 549 | 116 | |
XM_005265024.4 | XP_005265081.1 | 2 - 3 | 549 | 116 | |
XM_006713086.3 | XP_006713149.1 | 2 - 3 | 1205 | 116 | |
XM_011533570.2 | XP_011531872.1 | 2 - 3 | 669 | 116 | |
XM_011533571.2 | XP_011531873.1 | 2 - 3 | 606 | 116 | |
XM_017006113.1 | XP_016861602.1 | 2 - 3 | 963 | 116 | |
XM_017006114.1 | XP_016861603.1 | 2 - 3 | 1205 | 116 | |
XM_017006115.1 | XP_016861604.1 | 2 - 3 | 606 | 116 | |
XM_017006116.1 | XP_016861605.1 | 2 - 3 | 592 | 116 | |
XM_017006117.1 | XP_016861606.1 | 2 - 3 | 963 | 116 | |
XM_017006118.1 | XP_016861607.1 | 2 - 3 | 1205 | 116 | |
XM_017006119.1 | XP_016861608.1 | 2 - 3 | 592 | 116 | |
GenBank mRNA | AK001680.1 | - | 2 - 3 | 365 | 116 |
AK023263.1 | - | 2 - 3 | 587 | 116 | |
BC001622.1 | - | 2 - 3 | 380 | 116 | |
BC010421.1 | - | 2 - 3 | 937 | 116 |