The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015270.4 | NP_056085.1 | 16 - 17 | 2820 | 79 |
XM_006719210.3 | XP_006719273.1 | 16 - 17 | 3778 | 79 | |
XM_017018743.1 | XP_016874232.1 | 12 - 13 | 2982 | 79 | |
GenBank mRNA | AB007882.2 | - | 15 - 16 | 2602 | 79 |
AF250226.1 | - | 15 - 16 | 3319 | 79 | |
AK294446.1 | - | 3 - 4 | 815 | 79 | |
BC064923.1 | - | 17 - 18 | 2742 | 79 |