The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206942.1 | NP_001193871.1 | 4 - 5 | 1707 | 83 |
NM_006574.3 | NP_006565.2 | 4 - 5 | 1629 | 83 | |
GenBank mRNA | AF059274.1 | - | 4 - 5 | 1459 | 83 |
AF461087.1 | - | 4 - 5 | 3552 | 83 | |
AF461088.1 | - | 3 - 4 | 1561 | 83 | |
BM674119.1 | - | 1 - 2 | 83 | ||
BU732090.1 | - | 1 - 2 | 83 |