Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTX2
Entrez Gene ID
10651
Gene Name
metaxin 2
Gene Aliases
-
Chromosome Location
Chr.2: 176269395 - 176338025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006635.2 NP_001006636.1 5 - 6 645 121
NM_001319097.1 NP_001306026.1 4 - 5 441 121
NM_001319098.1 NP_001306027.1 4 - 5 441 121
NM_006554.4 NP_006545.1 4 - 5 441 121
XM_011510497.2 XP_011508799.1 5 - 6 538 121
GenBank mRNA AF053551.1 - 4 - 5 414 121
AK001287.1 - 4 - 5 393 121
AK289359.1 - 4 - 5 392 121
AK303564.1 - 5 - 6 570 121
AK315918.1 - 5 - 6 581 121
BC017271.2 - 5 - 6 575 121
BC067831.1 - 4 - 5 420 121
BC088359.1 - 4 - 5 376 121
BU165147.1 - 4 - 5 372 121
CX165048.1 - 4 - 5 334 121

View Full Product Details