The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006635.2 | NP_001006636.1 | 6 - 7 | 729 | 150 |
NM_001319097.1 | NP_001306026.1 | 5 - 6 | 525 | 150 | |
NM_001319098.1 | NP_001306027.1 | 5 - 6 | 525 | 150 | |
NM_006554.4 | NP_006545.1 | 5 - 6 | 525 | 150 | |
XM_011510497.2 | XP_011508799.1 | 6 - 7 | 622 | 150 | |
GenBank mRNA | AF053551.1 | - | 5 - 6 | 498 | 150 |
AK001287.1 | - | 5 - 6 | 477 | 150 | |
AK289359.1 | - | 5 - 6 | 476 | 150 | |
AK303564.1 | - | 6 - 7 | 654 | 150 | |
AK315918.1 | - | 6 - 7 | 665 | 150 | |
BC017271.2 | - | 6 - 7 | 659 | 150 | |
BC067831.1 | - | 5 - 6 | 504 | 150 | |
BC088359.1 | - | 5 - 6 | 460 | 150 | |
BU165147.1 | - | 5 - 6 | 456 | 150 | |
CX165048.1 | - | 5 - 6 | 418 | 150 |