Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR12
Entrez Gene ID
55759
Gene Name
WD repeat domain 12
Gene Aliases
YTM1
Chromosome Location
Chr.2: 202876958 - 202912226 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018256.3 NP_060726.3 10 - 11 89
XM_011511469.2 XP_011509771.1 9 - 10 89
GenBank mRNA AK001743.1 - 10 - 11 89
AK022781.1 - 10 - 11 89
AK022782.1 - 10 - 11 89
AK056092.1 - 10 - 11 89
AK299489.1 - 7 - 8 89
BC008082.2 - 10 - 11 89
BU633688.1 - 3 - 4 563 89
EF011617.1 - 10 - 11 89

View Full Product Details