The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270998.1 | NP_001257927.1 | 7 - 8 | 664 | 57 |
NM_001270999.1 | NP_001257928.1 | 6 - 7 | 535 | 57 | |
NM_138802.2 | NP_620157.1 | 6 - 7 | 788 | 57 | |
XM_006712285.1 | XP_006712348.1 | 7 - 8 | 641 | 57 | |
XM_006712286.3 | XP_006712349.1 | 7 - 8 | 667 | 57 | |
XM_017003368.1 | XP_016858857.1 | 7 - 8 | 667 | 57 | |
GenBank mRNA | AK074571.1 | - | 5 - 6 | 627 | 57 |
BC018415.1 | - | 6 - 7 | 756 | 57 | |
BU195818.1 | - | 7 - 8 | 619 | 57 |