Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAM2
Entrez Gene ID
10254
Gene Name
signal transducing adaptor molecule 2
Gene Aliases
Hbp, STAM2A, STAM2B
Chromosome Location
Chr.2: 152116801 - 152175992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005843.5 NP_005834.4 3 - 4 553 144
GenBank mRNA AF042273.1 - 3 - 4 312 144
AF042274.1 - 3 - 4 312 144
AK292265.1 - 3 - 4 248 144
AK292847.1 - 3 - 4 290 144
AK310863.1 - 3 - 4 290 144
BC028740.2 - 3 - 4 553 144

View Full Product Details