Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGM7
Entrez Gene ID
116179
Gene Name
transglutaminase 7
Gene Aliases
TGMZ
Chromosome Location
Chr.15: 43276277 - 43302255 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052955.2 NP_443187.1 4 - 5 689 72
XM_017021903.1 XP_016877392.1 4 - 5 687 72
GenBank mRNA AF363393.1 - 4 - 5 689 72

View Full Product Details