The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003246.3 | NP_003237.2 | 12 - 13 | 2103 | 98 |
XM_011521971.1 | XP_011520273.1 | 11 - 12 | 1925 | 98 | |
GenBank mRNA | AB209912.1 | - | 12 - 13 | 2089 | 98 |
AK291639.1 | - | 12 - 13 | 2103 | 98 | |
AK304754.1 | - | 12 - 13 | 1849 | 98 | |
AK310820.1 | - | 2 - 3 | 638 | 98 | |
BC136469.1 | - | 12 - 13 | 2077 | 98 | |
BC136470.1 | - | 12 - 13 | 2077 | 98 | |
X04665.1 | - | 12 - 13 | 1999 | 98 | |
X14787.1 | - | 12 - 13 | 2035 | 98 |