Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THBS1
Entrez Gene ID
7057
Gene Name
thrombospondin 1
Gene Aliases
THBS, THBS-1, TSP, TSP-1, TSP1
Chromosome Location
Chr.15: 39581079 - 39598918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003246.3 NP_003237.2 3 - 4 804 84
XM_011521971.1 XP_011520273.1 3 - 4 800 84
GenBank mRNA AB209912.1 - 3 - 4 790 84
AK291639.1 - 3 - 4 804 84
AK304754.1 - 3 - 4 550 84
BC028145.1 - 2 - 3 659 84
BC136469.1 - 3 - 4 778 84
BC136470.1 - 3 - 4 778 84
M14326.1 - 3 - 4 745 84
M25631.1 - 3 - 4 724 84
X04665.1 - 3 - 4 700 84
X14787.1 - 3 - 4 736 84

View Full Product Details