The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003246.3 | NP_003237.2 | 5 - 6 | 1085 | 78 |
XM_011521971.1 | XP_011520273.1 | 5 - 6 | 1081 | 78 | |
GenBank mRNA | AB209912.1 | - | 5 - 6 | 1071 | 78 |
AK291639.1 | - | 5 - 6 | 1085 | 78 | |
AK304754.1 | - | 5 - 6 | 831 | 78 | |
BC028145.1 | - | 4 - 5 | 940 | 78 | |
BC136469.1 | - | 5 - 6 | 1059 | 78 | |
BC136470.1 | - | 5 - 6 | 1059 | 78 | |
M14326.1 | - | 5 - 6 | 1026 | 78 | |
M25631.1 | - | 5 - 6 | 1005 | 78 | |
X04665.1 | - | 5 - 6 | 981 | 78 | |
X14787.1 | - | 5 - 6 | 1017 | 78 |