The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172779.1 | NP_001166250.1 | 8 - 9 | 1146 | 61 |
NM_001172780.1 | NP_001166251.1 | 8 - 9 | 1146 | 61 | |
NM_153353.4 | NP_699184.2 | 7 - 8 | 1050 | 61 | |
XM_006713508.3 | XP_006713571.1 | 8 - 9 | 876 | 61 | |
XM_011512442.2 | XP_011510744.1 | 8 - 9 | 927 | 61 | |
XM_017005746.1 | XP_016861235.1 | 8 - 9 | 1101 | 61 | |
GenBank mRNA | AK095125.1 | - | 8 - 9 | 891 | 61 |
AK130856.1 | - | 1 - 2 | 119 | 61 | |
AK293661.1 | - | 8 - 9 | 1024 | 61 | |
AK295065.1 | - | 8 - 9 | 885 | 61 | |
AK302466.1 | - | 8 - 9 | 908 | 61 | |
BC032841.2 | - | 7 - 8 | 789 | 61 |