Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
Chromosome Location
Chr.1: 224928422 - 225399294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373.1 NP_001364.1 65 - 66 10248 66
XM_006711735.3 XP_006711798.1 66 - 67 11665 66
XM_011544057.2 XP_011542359.1 67 - 68 11758 66
XM_011544058.2 XP_011542360.1 67 - 68 11755 66
XM_011544059.2 XP_011542361.1 67 - 68 11710 66
XM_011544061.2 XP_011542363.1 67 - 68 11653 66
XM_011544062.2 XP_011542364.1 67 - 68 11758 66
XM_011544063.2 XP_011542365.1 66 - 67 11599 66
XM_011544064.2 XP_011542366.1 66 - 67 11590 66
XM_011544065.2 XP_011542367.1 66 - 67 11566 66
XM_011544066.2 XP_011542368.1 66 - 67 11542 66
XM_011544067.2 XP_011542369.1 67 - 68 11758 66
XM_011544069.2 XP_011542371.1 65 - 66 11404 66
XM_011544070.2 XP_011542372.1 65 - 66 11380 66
XM_011544071.2 XP_011542373.1 64 - 65 11231 66
XM_011544072.2 XP_011542374.1 60 - 61 10441 66
XM_011544073.2 XP_011542375.1 54 - 55 11374 66
XM_011544074.2 XP_011542376.1 51 - 52 8611 66
XM_011544076.1 XP_011542378.1 49 - 50 7263 66
XM_011544079.1 XP_011542381.1 42 - 43 5996 66
XM_011544081.1 XP_011542383.1 33 - 34 4946 66
XM_017000295.1 XP_016855784.1 66 - 67 11683 66
XM_017000296.1 XP_016855785.1 67 - 68 11758 66
XM_017000297.1 XP_016855786.1 66 - 67 11641 66
XM_017000298.1 XP_016855787.1 67 - 68 11757 66
XM_017000299.1 XP_016855788.1 41 - 42 5842 66
GenBank mRNA CR936804.1 - 23 - 24 3209 66

View Full Product Details