Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
Chromosome Location
Chr.1: 224928422 - 225399294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373.1 NP_001364.1 75 - 76 12250 163
XM_006711735.3 XP_006711798.1 76 - 77 13667 163
XM_011544057.2 XP_011542359.1 77 - 78 13760 163
XM_011544058.2 XP_011542360.1 77 - 78 13757 163
XM_011544059.2 XP_011542361.1 77 - 78 13712 163
XM_011544061.2 XP_011542363.1 77 - 78 13655 163
XM_011544063.2 XP_011542365.1 76 - 77 13601 163
XM_011544064.2 XP_011542366.1 76 - 77 13592 163
XM_011544065.2 XP_011542367.1 76 - 77 13568 163
XM_011544066.2 XP_011542368.1 76 - 77 13544 163
XM_011544069.2 XP_011542371.1 75 - 76 13406 163
XM_011544070.2 XP_011542372.1 75 - 76 13382 163
XM_011544071.2 XP_011542373.1 74 - 75 13233 163
XM_011544072.2 XP_011542374.1 70 - 71 12443 163
XM_011544073.2 XP_011542375.1 64 - 65 13376 163
XM_011544074.2 XP_011542376.1 61 - 62 10613 163
XM_011544076.1 XP_011542378.1 59 - 60 9265 163
XM_011544079.1 XP_011542381.1 52 - 53 7998 163
XM_011544081.1 XP_011542383.1 43 - 44 6948 163
XM_017000295.1 XP_016855784.1 76 - 77 13685 163
XM_017000296.1 XP_016855785.1 77 - 78 13661 163
XM_017000297.1 XP_016855786.1 76 - 77 13643 163
XM_017000299.1 XP_016855788.1 51 - 52 7844 163

View Full Product Details