Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
Chromosome Location
Chr.1: 224928422 - 225399294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373.1 NP_001364.1 83 - 84 13544 71
XM_006711735.3 XP_006711798.1 84 - 85 14988 71
XM_011544057.2 XP_011542359.1 85 - 86 15081 71
XM_011544058.2 XP_011542360.1 85 - 86 15078 71
XM_011544059.2 XP_011542361.1 85 - 86 15033 71
XM_011544061.2 XP_011542363.1 85 - 86 14976 71
XM_011544062.2 XP_011542364.1 85 - 86 14925 71
XM_011544063.2 XP_011542365.1 84 - 85 14922 71
XM_011544064.2 XP_011542366.1 84 - 85 14913 71
XM_011544065.2 XP_011542367.1 84 - 85 14889 71
XM_011544066.2 XP_011542368.1 84 - 85 14865 71
XM_011544067.2 XP_011542369.1 84 - 85 14853 71
XM_011544069.2 XP_011542371.1 83 - 84 14727 71
XM_011544070.2 XP_011542372.1 83 - 84 14703 71
XM_011544071.2 XP_011542373.1 82 - 83 14554 71
XM_011544072.2 XP_011542374.1 78 - 79 13764 71
XM_011544073.2 XP_011542375.1 72 - 73 14697 71
XM_011544074.2 XP_011542376.1 69 - 70 11934 71
XM_011544076.1 XP_011542378.1 67 - 68 10586 71
XM_011544079.1 XP_011542381.1 60 - 61 9319 71
XM_011544081.1 XP_011542383.1 51 - 52 8269 71
XM_017000295.1 XP_016855784.1 84 - 85 15006 71
XM_017000296.1 XP_016855785.1 85 - 86 14982 71
XM_017000297.1 XP_016855786.1 84 - 85 14964 71
XM_017000299.1 XP_016855788.1 59 - 60 9165 71

View Full Product Details