Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTU2
Entrez Gene ID
348180
Gene Name
cytosolic thiouridylase subunit 2
Gene Aliases
C16orf84, NCS2, UPF0432
Chromosome Location
Chr.16: 88706463 - 88715386 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012759.2 NP_001012777.1 5 - 6 406 103
NM_001012762.2 NP_001012780.1 5 - 6 406 103
NM_001318513.1 NP_001305442.1 4 - 5 327 103
GenBank mRNA AK299521.1 - - 366 103
BC021056.1 - 5 - 6 350 103
BC080540.1 - 5 - 6 314 103
BC108659.1 - 5 - 6 354 103
BC121805.1 - 4 - 5 307 103
BC125269.1 - 5 - 6 386 103
BC157881.1 - 5 - 6 367 103
BI520274.1 - 5 - 6 406 103
BQ430809.1 - - 386 103

View Full Product Details