The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032856.3 | NP_116245.2 | 2 - 3 | 177 | 142 |
NR_130944.1 | 1 - 2 | 614 | 142 | ||
NR_130945.1 | 2 - 3 | 177 | 142 | ||
NR_130946.1 | 2 - 3 | 184 | 142 | ||
GenBank mRNA | AF161382.1 | - | 2 - 3 | 108 | 142 |
AK027794.1 | - | 2 - 3 | 114 | 142 | |
AK055578.1 | - | 1 - 2 | 614 | 142 | |
AK055729.1 | - | 2 - 3 | 114 | 142 | |
AK225068.1 | - | 2 - 3 | 117 | 142 | |
AK225199.1 | - | 2 - 3 | 114 | 142 | |
AK295372.1 | - | 2 - 3 | 108 | 142 | |
BC014115.2 | - | 2 - 3 | 115 | 142 | |
BC020252.2 | - | 1 - 2 | 66 | 142 | |
BC050648.1 | - | 2 - 3 | 134 | 142 | |
BC063392.1 | - | 2 - 3 | 145 | 142 | |
BU601517.1 | - | - | 187 | 142 | |
BX446522.2 | - | 2 - 3 | 107 | 142 | |
CN481562.1 | - | 2 - 3 | 113 | 142 |