The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032411.2 | NP_115787.1 | 1 - 2 | 185 | 85 |
GenBank mRNA | AF325503.1 | - | 1 - 2 | 185 | 85 |
AY358601.1 | - | 1 - 2 | 146 | 85 | |
BC021742.1 | - | 1 - 2 | 189 | 85 |