Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYL4
Entrez Gene ID
4635
Gene Name
myosin light chain 4
Gene Aliases
ALC1, AMLC, GT1, PRO1957
Chromosome Location
Chr.17: 47209017 - 47227650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002841.1 NP_001002841.1 7 - 8 740 79
NM_002476.2 NP_002467.1 6 - 7 687 79
XM_005257391.4 XP_005257448.1 7 - 8 667 79
XM_011524839.2 XP_011523141.2 7 - 8 827 79
XM_017024683.1 XP_016880172.1 7 - 8 814 79
GenBank mRNA AF116676.2 - - 1039 79
BC030228.1 - 6 - 7 676 79
M20641.1 - 3 - 4 303 79
M24121.1 - 6 - 7 674 79
M36172.1 - 6 - 7 668 79
X13955.1 - 6 - 7 668 79

View Full Product Details